Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Oxo-4-(1-pyrenyl)butyric acid ≥95%

Supplier: AMBEED, INC
Warning

A586420-1G A586420-25G A586420-5G
77627-212EA 15.45 USD InStock
77627-212 77627-214 77627-216
4-Oxo-4-(1-pyrenyl)butyric acid ≥95%
4-Oxo-4-(1-pyrenyl)butyric acid
Precaución: for Research Use Only


Formula: C₂₀H₁₄O₃
MDL Number: MFCD00010369
CAS Number: 7499-60-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty