Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Aminobutanenitrile hydrochloride ≥97%

Supplier: AMBEED, INC
Warning

A601325-1G A601325-250MG A601325-5G A601325-100MG
77747-241EA 809 USD InStock
77747-241 77747-242 77747-243 77747-240
4-Aminobutanenitrile hydrochloride ≥97%
4-Aminobutanenitrile hydrochloride
Precaución: for Research Use Only


Formula: C₄H₉ClN₂
MDL Number: MFCD16619402
CAS Number: 16011-90-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty