Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Piperazin-1-yl-2-phenylacetic acid, N4-BOC protected ≥95%

Supplier: AMBEED, INC
Warning

A626469-100MG A626469-1G A626469-250MG
77673-694EA 83 USD InStock
77673-694 77673-696 77673-698
2-Piperazin-1-yl-2-phenylacetic acid, N4-BOC protected ≥95%
2-Piperazin-1-yl-2-phenylacetic acid, N4-BOC protected
Precaución: for Research Use Only


Formula: C₁₇H₂₄N₂O₄
MW: 320.383 g/mol
MDL Number: MFCD01862496
CAS Number: 347186-49-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty