Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-3-(6-(3-(Difluoromethoxy)-5-fluorophenyl)-4-((3-(trifluoromethyl)phenyl)sulfonyl)-3,4-dihydro-2H-benzo[b][1,4]oxazin-2-yl)-2,2-dimethylpropanoic acid ≥99%

Supplier: AMBEED, INC
Warning

A627902-10MG A627902-250MG A627902-50MG
77617-644EA 65.92 USD InStock
77617-644 77749-280 77749-281
(S)-3-(6-(3-(Difluoromethoxy)-5-fluorophenyl)-4-((3-(trifluoromethyl)phenyl)sulfonyl)-3,4-dihydro-2H-benzo[b][1,4]oxazin-2-yl)-2,2-dimethylpropanoic acid ≥99%
(S)-3-(6-(3-(Difluoromethoxy)-5-fluorophenyl)-4-((3-(trifluoromethyl)phenyl)sulfonyl)-3,4-dihydro-2H-benzo[b][1,4]oxazin-2-yl)-2,2-dimethylpropanoic acid
Precaución: for Research Use Only


Formula: C₂₇H₂₃F₆NO₆S
MW: 603.53 g/mol
MDL Number: MFCD31544352
CAS Number: 2055536-64-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty