Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

HA 130 ≥98%

Supplier: AMBEED, INC
Warning

A662242-10MG A662242-5MG
77747-350EA 68 USD InStock
77747-350 77747-351
HA 130 ≥98%
HA 130
Precaución: for Research Use Only


Formula: C₂₄H₁₉BFNO₅S
MDL Number: MFCD20926343
CAS Number: 1229652-21-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty