Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cobalt (III) acetylacetonate ≥97%

Supplier: AMBEED, INC
Danger

A677284-10G A677284-25G A677284-5G
77595-034EA 8.2 USD InStock
77595-034 77602-722 77627-376
Cobalt (III) acetylacetonate ≥97%
Cobalt (III) acetylacetonate
Precaución: for Research Use Only


Formula: C₁₅H₂₁CoO₆
MDL Number: MFCD00013488
CAS Number: 21679-46-9
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty