Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Perylene Red ≥95%

Supplier: AMBEED, INC
Warning

A683460-250MG A683460-1G
77747-381EA 18 USD InStock
77747-381 77747-380
Perylene Red ≥95%
Perylene Red
Precaución: for Research Use Only


Formula: C₇₂H₅₈N₂O₈
MW: 1079.26 g/mol
MDL Number: MFCD21608638
CAS Number: 123174-58-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty