Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Adenosine 5-diphosphoribose sodium salt ≥98%

Supplier: AMBEED, INC
Warning

A685257-10MG A685257-5MG
77627-388EA 92.7 USD InStock
77627-388 77627-390
Adenosine 5-diphosphoribose sodium salt ≥98%
Adenosine 5-diphosphoribose sodium salt
Precaución: for Research Use Only


Formula: C₁₅H₂₂N₅NaO₁₄P₂
MW: 581.3 g/mol
MDL Number: MFCD00149359
CAS Number: 68414-18-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty