Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyl isothiocyanate ≥98%

Supplier: AMBEED, INC
Danger

A690663-1G A690663-25G A690663-5G
77614-132EA 11.33 USD InStock
77614-132 77686-669 77686-670
Benzyl isothiocyanate ≥98%
Benzyl isothiocyanate
Precaución: for Research Use Only


Formula: C₈H₇NS
MW: 149.22 g/mol
Boiling Pt: 243
Melting Pt: 24
MDL Number: MFCD00004819
CAS Number: 622-78-6
UN: 2810
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty