Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Palmatine chloride ≥98%

Supplier: AMBEED, INC
Warning

A697812-100G A697812-25G
77669-880EA 142 USD InStock
77669-880 77669-882
Palmatine chloride ≥98%
Palmatine chloride
Precaución: for Research Use Only


Formula: C₂₁H₂₂ClNO₄
MW: 387.86 g/mol
MDL Number: MFCD00016656
CAS Number: 10605-02-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty