Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-[7-(tert-Butyl)pyren-2-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98% (by GC, titration analysis)

Supplier: AMBEED, INC
Warning

A702923-100MG A702923-250MG
77622-504EA 101.97 USD InStock
77622-504 77622-506
2-[7-(tert-Butyl)pyren-2-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98% (by GC, titration analysis)
2-[7-(tert-Butyl)pyren-2-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only


Formula: C₂₆H₂₉BO₂
MDL Number: MFCD31618106
CAS Number: 1270030-08-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty