Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Bromoethoxy)trimethylsilane ≥95%

Supplier: AMBEED, INC
Danger

A705841-250MG A705841-1G A705841-5G
77749-447EA 98 USD InStock
77749-447 77749-446 77749-448
(2-Bromoethoxy)trimethylsilane ≥95%
(2-Bromoethoxy)trimethylsilane
Precaución: for Research Use Only


Formula: C₅H₁₃BrOSi
MW: 197.15 g/mol
CAS Number: 34714-03-9
UN: 1993
ADR: 3 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty