Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ACY-775 ≥98%

Supplier: AMBEED, INC
Danger

A725076-1MG A725076-50MG
77622-606EA 48.41 USD InStock
77622-606 77622-608
ACY-775 ≥98%
ACY-775


Formula: C₁₇H₁₉FN₄O₂
MDL Number: MFCD30533691
CAS Number: 1375466-18-4
UN: 1325
ADR: 4.1,Ⅲ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty