Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N-Dimethyl-N-(4-(2-(p-tolyl)-6,7-dihydro-5H-benzo[7]annulene-8-carboxamido)benzyl)tetrahydro-2H-pyran-4-aminium chloride ≥98%

Supplier: AMBEED, INC
Warning

A734428-10MG A734428-5MG
77686-737EA 98 USD InStock
77686-737 77686-738
N,N-Dimethyl-N-(4-(2-(p-tolyl)-6,7-dihydro-5H-benzo[7]annulene-8-carboxamido)benzyl)tetrahydro-2H-pyran-4-aminium chloride ≥98%
N,N-Dimethyl-N-(4-(2-(p-tolyl)-6,7-dihydro-5H-benzo[7]annulene-8-carboxamido)benzyl)tetrahydro-2H-pyran-4-aminium chloride
Precaución: for Research Use Only


Formula: C₃₃H₃₉ClN₂O₂
MDL Number: MFCD05662319
CAS Number: 229005-80-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty