Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-((4-Oxo-3-phenyl-3,4,6,7-tetrahydrothieno[3,2-d]pyrimidin-2-yl)thio)-N-(5-phenylpyridin-2-yl)acetamide ≥98%

Supplier: AMBEED, INC
Warning

A735189-10MG A735189-5MG
77686-742EA 66 USD InStock
77686-742 77686-743
2-((4-Oxo-3-phenyl-3,4,6,7-tetrahydrothieno[3,2-d]pyrimidin-2-yl)thio)-N-(5-phenylpyridin-2-yl)acetamide ≥98%
2-((4-Oxo-3-phenyl-3,4,6,7-tetrahydrothieno[3,2-d]pyrimidin-2-yl)thio)-N-(5-phenylpyridin-2-yl)acetamide
Precaución: for Research Use Only


Formula: C₂₅H₂₀N₄O₂S₂
MDL Number: MFCD27937771
CAS Number: 1427782-89-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty