Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 3-hydroxypropionate ≥98%

Supplier: AMBEED, INC
Warning

A736070-100MG A736070-1G A736070-5G
77674-016EA 40 USD InStock
77674-016 77674-018 77674-020
tert-Butyl 3-hydroxypropionate ≥98%
tert-Butyl 3-hydroxypropionate
Precaución: for Research Use Only


Formula: C₇H₁₄O₃
MW: 146.184 g/mol
MDL Number: MFCD04974574
CAS Number: 59854-11-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty