Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(3-(4-Fluorophenyl)-1-phenyl-1H-pyrazol-4-yl)imidazolidine-2,4-dione ≥98%

Supplier: AMBEED, INC
Warning

A749014-10MG A749014-5MG
77686-755EA 95 USD InStock
77686-755 77686-756
5-(3-(4-Fluorophenyl)-1-phenyl-1H-pyrazol-4-yl)imidazolidine-2,4-dione ≥98%
5-(3-(4-Fluorophenyl)-1-phenyl-1H-pyrazol-4-yl)imidazolidine-2,4-dione
Precaución: for Research Use Only


Formula: C₁₈H₁₃FN₄O₂
MDL Number: MFCD03268950
CAS Number: 484049-04-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty