Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Pyridin-2-yl)-1H-pyrrole-3-carboxylic acid ≥98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A751871-1G A751871-250MG
77684-682EA 105 USD InStock
77684-682 77684-683
4-(Pyridin-2-yl)-1H-pyrrole-3-carboxylic acid ≥98%, Ambeed.Inc
4-(Pyridin-2-yl)-1H-pyrrole-3-carboxylic acid
Precaución: For research use only.


Formula: C₁₀H₈N₂O₂
MDL Number: MFCD11217989
CAS Number: 1096815-73-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty