Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Mal-amido-PEG6-acid ≥95%

Supplier: AMBEED, INC
Warning

A754937-100MG A754937-250MG
77622-772EA 51.5 USD InStock
77622-772 77622-774
Mal-amido-PEG6-acid ≥95%
Mal-amido-PEG6-acid
Precaución: for Research Use Only


Formula: C₂₂H₃₆N₂O₁₁
MDL Number: MFCD21363276
CAS Number: 1334177-79-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty