Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(11aR)-3,7-Bis[3,5-bis(trifluoromethyl)phenyl]-10,11,12,13-tetrahydro-5-hydroxy-5-oxide-diindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin ≥98%

Supplier: AMBEED, INC
Warning

A759881-250MG A759881-50MG A759881-1G
77747-499EA 436 USD InStock
77747-499 77747-500 77747-498
(11aR)-3,7-Bis[3,5-bis(trifluoromethyl)phenyl]-10,11,12,13-tetrahydro-5-hydroxy-5-oxide-diindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin ≥98%
(11aR)-3,7-Bis[3,5-bis(trifluoromethyl)phenyl]-10,11,12,13-tetrahydro-5-hydroxy-5-oxide-diindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin
Precaución: for Research Use Only


Formula: C₃₃H₁₉F₁₂O₄P
MDL Number: MFCD29905018
CAS Number: 1297613-76-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty