Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

TCO-pnb ester 95% rel-(1R-4E-pS)-axial

Supplier: AMBEED, INC
Warning

A760780-1G A760780-250MG
77622-868EA 1071.2 USD InStock
77622-868 77622-870
TCO-pnb ester 95% rel-(1R-4E-pS)-axial
TCO-pnb ester
Precaución: for Research Use Only


Formula: C₁₅H₁₇NO₅
MDL Number: MFCD29052196
CAS Number: 2389122-16-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty