Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-4-hydroxy-D-phenylglycine ≥95%

Supplier: AMBEED, INC
Warning

A765141-25G A765141-5G
77686-775EA 309 USD InStock
77686-775 77686-776
Fmoc-4-hydroxy-D-phenylglycine ≥95%
Fmoc-4-hydroxy-D-phenylglycine
Precaución: for Research Use Only


Formula: C₂₃H₁₉NO₅
MDL Number: MFCD02682475
CAS Number: 178119-93-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty