Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Cyanomethyl)-5-fluorobenzeneboronic acid ≥98%

Supplier: AMBEED, INC
Warning

A788941-250MG A788941-1G
77747-535EA 187 USD InStock
77747-535 77747-534
3-(Cyanomethyl)-5-fluorobenzeneboronic acid ≥98%
3-(Cyanomethyl)-5-fluorobenzeneboronic acid
Precaución: for Research Use Only


Formula: C₈H₇BFNO₂
MDL Number: MFCD27935531
CAS Number: 1460307-63-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty