Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Chlorobenzyl)piperidine hydrochloride ≥95%

Supplier: AMBEED, INC
Warning

A818986-250MG A818986-1G
77674-208EA 113 USD InStock
77674-208 77674-206
4-(4-Chlorobenzyl)piperidine hydrochloride ≥95%
4-(4-Chlorobenzyl)piperidine hydrochloride


Formula: C₁₂H₁₆ClN·ClH
MDL Number: MFCD03840138
CAS Number: 36968-94-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty