Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[2-(2-Hydroxy-ethoxy)-ethyl]-carbamic acid 9H-fluoren-9-ylmethyl ester ≥98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A823086-1G A823086-5G
77684-777EA 46 USD InStock
77684-777 77684-778
[2-(2-Hydroxy-ethoxy)-ethyl]-carbamic acid 9H-fluoren-9-ylmethyl ester ≥98%, Ambeed.Inc
[2-(2-Hydroxy-ethoxy)-ethyl]-carbamic acid 9H-fluoren-9-ylmethyl ester
Precaución: For research use only.


Formula: C₁₉H₂₁NO₄
MDL Number: MFCD04974286
CAS Number: 299430-87-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty