Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methoxy-3-pyridineacetic acid ≥98%

Supplier: AMBEED, INC
Warning

A823718-1G A823718-5G
77674-216EA 39 USD InStock
77674-216 77674-218
6-Methoxy-3-pyridineacetic acid ≥98%
6-Methoxy-3-pyridineacetic acid
Precaución: for Research Use Only


Formula: C₈H₉NO₃
MDL Number: MFCD09924951
CAS Number: 902130-87-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty