Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)pyridine ≥98%

Supplier: AMBEED, INC
Warning

A834881-5G A834881-250MG A834881-100MG A834881-1G A834881-10G
77749-634EA 350 USD InStock
77749-634 77749-633 77749-630 77749-632 77749-631
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)pyridine ≥98%
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)pyridine
Precaución: for Research Use Only


Formula: C₁₂H₁₅BF₃NO₂
MW: 273.06 g/mol
MDL Number: MFCD13182147
CAS Number: 1383625-22-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty