Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl tricosanoate ≥95%

Supplier: AMBEED, INC
Warning

A849595-100MG A849595-250MG A849595-5G
77670-222EA 38 USD InStock
77670-222 77670-224 77670-226
Methyl tricosanoate ≥95%
Methyl tricosanoate
Precaución: for Research Use Only


Formula: C₂₄H₄₈O₂
MDL Number: MFCD00009349
CAS Number: 2433-97-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty