Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Oseltamivir acid ≥98%

Supplier: AMBEED, INC
Warning

A850094-100MG A850094-10MG A850094-5MG
77686-883EA 319 USD InStock
77686-883 77686-884 77686-885
Oseltamivir acid ≥98%
Oseltamivir acid
Precaución: for Research Use Only


Formula: C₁₄H₂₄N₂O₄
MDL Number: MFCD00953940
CAS Number: 187227-45-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty