Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L-Glutamic acid ammonium salt ≥98%

Supplier: AMBEED, INC
Warning

A858216-5G A858216-100G A858216-25G
77603-166EA 15.38 USD InStock
77603-166 77670-242 77670-244
L-Glutamic acid ammonium salt ≥98%
L-Glutamic acid ammonium salt


Formula: C₅H₈NO₄·NH₄
MDL Number: MFCD00039098
CAS Number: 7558-63-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty