Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

CP-547632 ≥95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A867793-10MG A867793-5MG
77684-829EA 144 USD InStock
77684-829 77684-830
CP-547632 ≥95%, Ambeed.Inc
CP-547632
Precaución: For research use only.


Formula: C₂₀H₂₄BrF₂N₅O₃S
MDL Number: MFCD07772279
CAS Number: 252003-65-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty