Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Rifabutin ≥96%

Supplier: AMBEED, INC
Warning

A878035-100MG A878035-1G A878035-250MG
77670-268EA 32 USD InStock
77670-268 77670-270 77670-272
Rifabutin ≥96%
Rifabutin
Precaución: for Research Use Only


Formula: C₄₆H₆₂N₄O₁₁
MDL Number: MFCD00866816
CAS Number: 72559-06-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty