Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Oxoheptanoic acid ≥97%

Supplier: AMBEED, INC
Warning

A903727-100MG A903727-1G
77749-716EA 236 USD InStock
77749-716 77749-717
7-Oxoheptanoic acid ≥97%
7-Oxoheptanoic acid
Precaución: for Research Use Only


Formula: C₇H₁₂O₃
MW: 144.17 g/mol
MDL Number: MFCD00047662
CAS Number: 35923-65-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty