Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

RGB-286638 ≥98%

Supplier: AMBEED, INC
Warning

A908306-5MG A908306-10MG
77747-671EA 74 USD InStock
77747-671 77747-670
RGB-286638 ≥98%
RGB-286638
Precaución: for Research Use Only


Formula: C₂₉H₃₅N₇O₄
MDL Number: MFCD18633230
CAS Number: 784210-88-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty