Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-butyl 3-hydroxy-3-(hydroxymethyl)azetidine-1-carboxylate ≥98%

Supplier: AMBEED, INC
Warning

A916476-100MG A916476-1G A916476-250MG
77749-739EA 122 USD InStock
77749-739 77749-740 77749-741
tert-butyl 3-hydroxy-3-(hydroxymethyl)azetidine-1-carboxylate ≥98%
tert-butyl 3-hydroxy-3-(hydroxymethyl)azetidine-1-carboxylate
Precaución: for Research Use Only


Formula: C₉H₁₇NO₄
MDL Number: MFCD30724024
CAS Number: 1428330-70-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty