Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Aminoindole hydrochloride ≥98%

Supplier: AMBEED, INC
Warning

A916631-1G A916631-250MG
77749-742EA 406 USD InStock
77749-742 77749-743
3-Aminoindole hydrochloride ≥98%
3-Aminoindole hydrochloride
Precaución: for Research Use Only


Formula: C₈H₈N₂·ClH
MDL Number: MFCD09800384
CAS Number: 57778-93-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty