Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α,α-Diphenyl-N-methyl-L-prolinol ≥97%

Supplier: AMBEED, INC
Warning

A946252-25G A946252-5G A946252-1G A946252-250MG
77674-408EA 78 USD InStock
77674-408 77674-410 77674-404 77674-406
α,α-Diphenyl-N-methyl-L-prolinol ≥97%
α,α-Diphenyl-N-methyl-L-prolinol
Precaución: for Research Use Only


Formula: C₁₈H₂₁NO
MDL Number: MFCD00145245
CAS Number: 110529-22-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty