Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-((7-(3,4-Dimethoxyphenyl)imidazo[1,2-c]pyrimidin-5-yl)amino)nicotinamide ≥99%

Supplier: AMBEED, INC
Warning

A954369-5MG A954369-10MG
77747-712EA 71 USD InStock
77747-712 77747-711
2-((7-(3,4-Dimethoxyphenyl)imidazo[1,2-c]pyrimidin-5-yl)amino)nicotinamide ≥99%
2-((7-(3,4-Dimethoxyphenyl)imidazo[1,2-c]pyrimidin-5-yl)amino)nicotinamide
Precaución: for Research Use Only


Formula: C₂₀H₁₈N₆O₃
MDL Number: MFCD18641826
CAS Number: 732983-37-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty