Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-((1S,2R,4S,5R)-5-ethylquinuclidin-2-yl)(6-methoxyquinolin-4-yl)methanamine trihydrochloride ≥97%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A981506-100MG A981506-250MG
77684-908EA 38 USD InStock
77684-908 77684-909
(R)-((1S,2R,4S,5R)-5-ethylquinuclidin-2-yl)(6-methoxyquinolin-4-yl)methanamine trihydrochloride ≥97%, Ambeed.Inc
(R)-((1S,2R,4S,5R)-5-ethylquinuclidin-2-yl)(6-methoxyquinolin-4-yl)methanamine trihydrochloride
Precaución: For research use only.


Formula: C₂₀H₃₀Cl₃N₃O
MDL Number: MFCD22200492
CAS Number: 931098-92-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty