Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N'-(2-Fluoro-5-(pyridazin-4-yl)benzoyl)benzenesulfonohydrazide 99+%

Supplier: AMBEED, INC
Warning

A983022-50MG A983022-100MG
77749-822EA 144 USD InStock
77749-822 77749-821
N'-(2-Fluoro-5-(pyridazin-4-yl)benzoyl)benzenesulfonohydrazide 99+%
N'-(2-Fluoro-5-(pyridazin-4-yl)benzoyl)benzenesulfonohydrazide
Precaución: for Research Use Only


Formula: C₁₇H₁₃FN₄O₃S
MDL Number: MFCD31617713
CAS Number: 2055397-88-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty