Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-L-HoLys(Boc)-OH ≥97%

Supplier: AMBEED, INC
Warning

A996060-1G A996060-100MG
77603-406EA 479.7 USD InStock
77603-406 77670-446
Fmoc-L-HoLys(Boc)-OH ≥97%
Fmoc-L-HoLys(Boc)-OH


Formula: C₂₇H₃₄N₂O₆
MDL Number: MFCD01860606
CAS Number: 194718-17-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty