Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Propargyl-PEG8-amine ≥97%

Supplier: AMBEED, INC
Warning

A997135-1G A997135-250MG
77749-831EA 281 USD InStock
77749-831 77749-832
Propargyl-PEG8-amine ≥97%
Propargyl-PEG8-amine
Precaución: for Research Use Only


Formula: C₁₉H₃₇NO₈
MDL Number: MFCD28334507
CAS Number: 1196732-52-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty