Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl (1-diazo-2-oxopropyl)phosphonate 10% in acetonitrile

Supplier: Decon Laboratories Ltd
Danger

D5048-25G D5048-5G
TCD5048-25GEA 307 USD InStock
TCD5048-25G TCD5048-5G
Dimethyl (1-diazo-2-oxopropyl)phosphonate 10% in acetonitrile
Dimethyl (1-diazo-2-oxopropyl)phosphonate


Formula: C₅H₉N₂O₄P
MW: 192.11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07368360
CAS Number: 90965-06-3
UN: 1648
ADR: 3 II
Merck Index: 14,01177
Order Now
Specification Test Results
Appearance Light yellow to Yellow to Orange clear liquid
Concentration (HNMR) 10.0 to 11.0 w/w%

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty