Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium 2,4,8,10-Tetra-tert-butyl-12H-dibenzo[d,g][1,3,2]dioxaphosphocin-6-olate 6-Oxide ≥98.0% (by HPLC, titration analysis)

Supplier: Decon Laboratories Ltd
Warning

O0527-25G O0527-5G
TCO0527-25GEA 129 USD InStock
TCO0527-25G TCO0527-5G
Sodium 2,4,8,10-Tetra-tert-butyl-12H-dibenzo[d,g][1,3,2]dioxaphosphocin-6-olate 6-Oxide ≥98.0% (by HPLC, titration analysis)
Sodium 2,4,8,10-Tetra-tert-butyl-12H-dibenzo[d,g][1,3,2]dioxaphosphocin-6-olate 6-Oxide


Formula: C₂₉H₄₂NaO₄P
MDL Number: MFCD01748150
CAS Number: 85209-91-2
EINECS: 286-344-4
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty