Brady® Aggressive Adhesive Multi-Purpose Polyester Labels
Supplier: Brady Worldwide
170732 170733 170734 170735 170736 170737 170738 170739 170859 170730 170731
76796-142EA 60.28 USD InStock
76796-142 76795-868 76796-128 76796-060 76795-948 76795-780 76796-180 76795-974 76796-168 76796-148 76795-940
Brady® Aggressive Adhesive Multi-Purpose Polyester Labels
Labels
Versatile labels are designed for a wide range of applications such as labeling equipment, circuit boards, panel components, and barcodes.
  • Permanent acrylic adhesive
  • Gloss finish
  • Chemical-resistant, corrosion-resistant, humidity-resistant, UV-resistant, weather-resistant material
  • Recommended ribbon: R6000HF
Constructed to withstand abrasions, extreme temperatures, and solvents such as alcohol, oil and water. Made with a permanent acrylic adhesive that sticks to rough, smooth, and powder-coated flat surfaces.
Certificaciones: UL recognized to UL969, CSA to C22.2 No.0.15-95, RoHS compliant to 2011/65/EU (Pass 2015/863).
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty