HyFlex® 11-591 Industrial Gloves
Supplier: Ansell Healthcare
11591060 11591070 11591080 11591090 11591100 11591110 11591120
77322-776CS 1996.79 USD InStock
77322-776 77322-778 77322-780 77322-782 77322-784 77322-786 77322-788
HyFlex® 11-591 Industrial Gloves
Gloves
HyFlex® 11-591 industrial gloves provide enhanced comfort, with a 20% lighter design that allows extended use on varied tasks.
  • Ultra-lightweight design
  • 12 times greater cut resistance for reliable hand protection
  • Palm dipped, thumbcrotch reinforcement
  • Silicone-free and anti-static
HyFlex® 11-591 industrial gloves offer 12 times greater cut resistance thanks to the Next Generation single HPPE yarn developed with INTERCEPT™ Cut Resistance Technology for reliable hand protection. Featuring a 20% lighter design, these gloves provide enhanced comfort, and allow workers to perform varied tasks while keeping hands protected throughout extended use. Improved FORTIX™ Technology delivers up to 20% more abrasion resistance for longer-lasting handling in abrasive conditions. EN 407 contact heat level 1 to protect hands while handling hot parts.
HyFlex® 11-591 industrial gloves are recommended for use in automotive, glass manufacturing, heavy manufacturing, metal fabrication, machinery, equipment, and pulp/paper industries.
Certificaciones: ANSI Cut A9. ANSI ABR 5. EN 388:2016 +A1:2018, EN407:2020, EN ISO 21420:2020. Food Contact approved. Category II.
Envase: Gloves are packaged 12 pairs per paper band bundle, 12 paper band bundles per carton.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty