Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Acetonitrile 99.6% ACS

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

423250010 423250025 423250100 423255000
TS423250010EA 135 USD InStock
TS423250010 TS423250025 TS423250100 TS423255000
Acetonitrile 99.6% ACS
Acetonitrile


Formula: H₃CCN
Boiling Pt: 81…82 °C (760 mmHg)
Melting Pt: –46 °C
Density: 0.781 g/cm³
Flash Pt: 2 °C
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Order Now
Specification Test Results
Appearance Clear liquid
Water ≤0.3 % (K.F.)
GC ≥99.5 %
Color scale ≤10 APHA
Residue after evaporation ≤0.005 %
Titrable acid ≤0.008 meq/g
Titrable base ≤0.0006 meq/g

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty