Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Acetone 99.9% (by GC, corrected for water content), ECD tested halocarbons free grade for residue analysis

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

326700010 326700025
TS326700010EA 60 USD InStock
TS326700010 TS326700025
Acetone 99.9% (by GC, corrected for water content), ECD tested halocarbons free grade for residue analysis
Acetone


Formula: CH₃COCH₃
Boiling Pt: 56.2 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.79 g/cm³
Flash Pt: –20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Order Now
Specification Test Results
Appearance Clear liquid
Water ≤0.2 % (Coulometric)
GC 99.9 to 100 % (on anhydrous basis)
Color scale ≤10 APHA
GC/ECD (as lindane) ≤2 ng/l (EOX suitab.)
GC/ECD ≤5 ng/l (volatile imp. (as E.Dibromide))
Residue after evaporation ≤0.0002 %
Acidity (CH3COOH) ≤0.002 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty