Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Acetone, meets analytical specification of Ph.Eur.

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

447450010 447450250
TS447450010EA 56 USD InStock
TS447450010 TS447450250
Acetone, meets analytical specification of Ph.Eur.
Acetone


Formula: CH₃COCH₃
Boiling Pt: 56 °C (760 mmHg)
Melting Pt: –95 °C
Density: 0.79 g/cm³
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
UN: 1090
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Water ≤0.3 %
Appearance of solution Clear colorless
Identification Passes test
Relative density 0.790 to 0.793
Residue after evaporation ≤50 ppm
Related substances ≤2 ppm (Benzene)
Acidity/alkalinity Passes test
Insoluble matter (in water) Passes test
Reducing substances Passes test

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty