Human recombinant Flt-3L-Ig
Supplier: BIO X CELL
BE0098-1MG BE0098-5MG
77578-784EA 464.15 USD InStock
77578-784 77578-790
Human recombinant Flt-3L-Ig
Proteins and Peptides
Flt-3L (FMS-related Tyrosine Kinase 3 Ligand) is an endogenous protein that functions as a cytokine and growth factor. Flt-3L is crucial for the development of conventional dendritic cells (cDCs) and plasmacytoid dendritic cells (pDCs). Recombinant Flt-3L-Ig is a fusion protein consisting of human Flt-3L fused to the Fc portion of human IgG1. This fusion protein is useful for activating Flt3 signaling and inducing the expansion of DC populations. Human Flt-3L-Ig is frequently reported to stimulate Flt3 signaling in vivo in mice.
  • Fusion Protein
  • Clone: Flt-3L-Ig (hum/hum)
  • Reactivities: Mouse, Human
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty